Anti-IREB2 Mouse Monoclonal Antibody [clone: 1F3]

Supplier: US Biological
USBI128578EA 702 CHF
USBI128578
Anti-IREB2 Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Type: Primary
Antigen: IREB2 (iron-responsive element binding protein 2)
Clonality: monoclonal
Clone: 1F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR