Anti-IRAK4 Mouse Monoclonal Antibody [clone: 2D3]

Supplier: US Biological
USBI128576EA 702 CHF
USBI128576
Anti-IRAK4 Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
Recommended dilution: Immunochemistry: 6?g/ml

Type: Primary
Antigen: IRAK4 (interleukin-1 receptor-associated kinase 4)
Clonality: monoclonal
Clone: 2D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR