Anti-IRAK3 Mouse Monoclonal Antibody [clone: 1F6]

Supplier: US Biological
USBI128575EA 702 CHF
USBI128575
Anti-IRAK3 Mouse Monoclonal Antibody [clone: 1F6]
Antibodies
Type: Primary
Antigen: IRAK3 (interleukin-1 receptor-associated kinase 3)
Clonality: monoclonal
Clone: 1F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR