Anti-IRAK1 Mouse Monoclonal Antibody [clone: 3A9]

Supplier: US Biological
USBI128571EA 702 CHF
USBI128571
Anti-IRAK1 Mouse Monoclonal Antibody [clone: 3A9]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: IRAK1 (interleukin-1 receptor-associated kinase 1)
Clonality: monoclonal
Clone: 3A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR