Anti-IQGAP3 Mouse Monoclonal Antibody [clone: 6E3]

Supplier: US Biological
USBI128567EA 702 CHF
USBI128567
Anti-IQGAP3 Mouse Monoclonal Antibody [clone: 6E3]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: IQGAP3 (IQ motif containing GTPase activating protein 3)
Clonality: monoclonal
Clone: 6000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR