Anti-IQGAP2 Mouse Monoclonal Antibody [clone: 1B10]

Supplier: US Biological
USBI128564EA 659 CHF
USBI128564
Anti-IQGAP2 Mouse Monoclonal Antibody [clone: 1B10]
Antibodies
Type: Primary
Antigen: IQGAP2 (IQ motif containing GTPase activating protein 2)
Clonality: monoclonal
Clone: 1B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR