Anti-IQGAP1 Mouse Monoclonal Antibody [clone: 2C5]

Supplier: US Biological
USBI128563EA 702 CHF
USBI128563
Anti-IQGAP1 Mouse Monoclonal Antibody [clone: 2C5]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: IQGAP1 (IQ motif containing GTPase activating protein 1)
Clonality: monoclonal
Clone: 2C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR