Anti-IQCH Mouse Monoclonal Antibody [clone: 4H3]
Lieferant: US Biological
USBI128562EA 733 CHF
USBI128562
Anti-IQCH Mouse Monoclonal Antibody [clone: 4H3]
Antikörper
Type: Primary
Antigen: IQCH (IQ motif containing H)
Clonality: monoclonal
Clone: 4H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human