Anti-IQCB1 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128560EA 702 CHF
USBI128560
Anti-IQCB1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IQCB1 (IQ motif containing B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR