Anti-IPR1 Mouse Monoclonal Antibody [clone: 8C8]

Supplier: US Biological
USBI128559EA 702 CHF
USBI128559
Anti-IPR1 Mouse Monoclonal Antibody [clone: 8C8]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IPR1
Clonality: monoclonal
Clone: 8C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR