Anti-IP6K2 Mouse Monoclonal Antibody [clone: 1C6]
Lieferant: US Biological
USBI128554EA 733 CHF
USBI128554
Anti-IP6K2 Mouse Monoclonal Antibody [clone: 1C6]
Antikörper
Type: Primary
Antigen: IP6K2 (inositol hexakisphosphate kinase 2)
Clonality: monoclonal
Clone: 1C6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human