Anti-IL31RA Mouse Monoclonal Antibody [clone: 3A10]

Supplier: US Biological
USBI128545EA 702 CHF
USBI128545
Anti-IL31RA Mouse Monoclonal Antibody [clone: 3A10]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IL31RA (interleukin 31 receptor A)
Clonality: monoclonal
Clone: 3A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR