Anti-IL24 Mouse Monoclonal Antibody [clone: 4B6]

Supplier: US Biological
USBI128543EA 702 CHF
USBI128543
Anti-IL24 Mouse Monoclonal Antibody [clone: 4B6]
Antibodies
Type: Primary
Antigen: IL24 (interleukin 24)
Clonality: monoclonal
Clone: 4B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR