Anti-Interleukin 18BP Mouse Monoclonal Antibody [clone: 2A9]

Lieferant: US Biological
USBI128539EA 733 CHF
USBI128539
Anti-Interleukin 18BP Mouse Monoclonal Antibody [clone: 2A9]
Antikörper
Type: Primary
Antigen: Interleukin 18BP
Clonality: monoclonal
Clone: 2A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR