Anti-IRF9 Mouse Monoclonal Antibody [clone: 1D11]

Supplier: US Biological
USBI128537EA 702 CHF
USBI128537
Anti-IRF9 Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IRF9 (interferon regulatory factor 9)
Clonality: monoclonal
Clone: 1D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR