Anti-INS Mouse Monoclonal Antibody [clone: 3E7]

Lieferant: US Biological
USBI128533EA 733 CHF
USBI128533
Anti-INS Mouse Monoclonal Antibody [clone: 3E7]
Antikörper
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone: 30000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR