Drucken

Anti-INPP5K Mouse Monoclonal Antibody [clone: 3B4]

Lieferant: US Biological
USBI128532EA 702 CHF
USBI128532
Anti-INPP5K Mouse Monoclonal Antibody [clone: 3B4]
Antikörper
Type: Primary
Antigen: INPP5K (inositol polyphosphate-5-phosphatase K)
Clonality: monoclonal
Clone: 3B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR