Anti-INPP5B Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128528EA 750 CHF
USBI128528
Anti-INPP5B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: INPP5B (inositol polyphosphate-5-phosphatase, 75kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR