Anti-INPP5A Mouse Monoclonal Antibody [clone: 3D8]

Supplier: US Biological
USBI128526EA 702 CHF
USBI128526
Anti-INPP5A Mouse Monoclonal Antibody [clone: 3D8]
Antibodies
Type: Primary
Antigen: INPP5A (inositol polyphosphate-5-phosphatase, 40kDa)
Clonality: monoclonal
Clone: 3D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR