Anti-INPP4B Mouse Monoclonal Antibody [clone: 3F2]

Supplier: US Biological
USBI128524EA 702 CHF
USBI128524
Anti-INPP4B Mouse Monoclonal Antibody [clone: 3F2]
Antibodies
Type: Primary
Antigen: INPP4B (inositol polyphosphate-4-phosphatase, type II, 105kDa)
Clonality: monoclonal
Clone: 3F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR