Anti-INHBE Mouse Monoclonal Antibody [clone: 4B5]

Lieferant: US Biological
USBI128519EA 733 CHF
USBI128519
Anti-INHBE Mouse Monoclonal Antibody [clone: 4B5]
Antikörper
Type: Primary
Antigen: INHBE (inhibin, beta E)
Clonality: monoclonal
Clone: 4B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR