Anti-INHBE Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128518EA 788 CHF
USBI128518
Anti-INHBE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: INHBE (inhibin, beta E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR