Anti-INHBB Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128515EA 750 CHF
USBI128515
Anti-INHBB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: INHBB (inhibin, beta B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR