Anti-ING3 Mouse Monoclonal Antibody [clone: 2C4]

Supplier: US Biological
USBI128509EA 702 CHF
USBI128509
Anti-ING3 Mouse Monoclonal Antibody [clone: 2C4]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ING3 (inhibitor of growth family, member 3)
Clonality: monoclonal
Clone: 2C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR