Anti-ING2 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: US Biological
USBI128508EA 702 CHF
USBI128508
Anti-ING2 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
Type: Primary
Antigen: ING2 (inhibitor of growth family, member 2)
Clonality: monoclonal
Clone: 1D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR