Anti-ING1 Mouse Monoclonal Antibody [clone: 2F9]

Supplier: US Biological
USBI128507EA 702 CHF
USBI128507
Anti-ING1 Mouse Monoclonal Antibody [clone: 2F9]
Antibodies
Type: Primary
Antigen: ING1 (inhibitor of growth family, member 1)
Clonality: monoclonal
Clone: 2F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR