Anti-ING1 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128506EA 788 CHF
USBI128506
Anti-ING1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ING1 (inhibitor of growth family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR