Anti-IDO1 Mouse Monoclonal Antibody [clone: 4D2]

Supplier: US Biological
USBI128503EA 702 CHF
USBI128503
Anti-IDO1 Mouse Monoclonal Antibody [clone: 4D2]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IDO1 (indoleamine 2,3-dioxygenase 1)
Clonality: monoclonal
Clone: 4D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR