Anti-IMPDH2 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128499EA 750 CHF
USBI128499
Anti-IMPDH2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IMPDH2 (IMP (inosine 5'-monophosphate) dehydrogenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR