Anti-IMPDH1 Mouse Monoclonal Antibody [clone: 3G6]

Supplier: US Biological
USBI128497EA 702 CHF
USBI128497
Anti-IMPDH1 Mouse Monoclonal Antibody [clone: 3G6]
Antibodies
Type: Primary
Antigen: IMPDH1 (IMP (inosine 5'-monophosphate) dehydrogenase 1)
Clonality: monoclonal
Clone: 3G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR