Anti-IMPACT Mouse Polyclonal Antibody

Supplier: US Biological
USBI128493EA 702 CHF
USBI128493
Anti-IMPACT Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IMPACT (impact RWD domain protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR