Anti-IMPA1 Mouse Monoclonal Antibody [clone: 1E6-F11]

USBI128491EA 659 CHF
USBI128491
Anti-IMPA1 Mouse Monoclonal Antibody [clone: 1E6-F11]
Antikörper
Type: Primary
Antigen: IMPA1 (inositol(myo)-1(or 4)-monophosphatase 1)
Clonality: monoclonal
Clone: 1E6-F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR