Anti-IMP3 Mouse Monoclonal Antibody [clone: 3B12]

Supplier: US Biological
USBI128489EA 702 CHF
USBI128489
Anti-IMP3 Mouse Monoclonal Antibody [clone: 3B12]
Antibodies
Type: Primary
Antigen: IMP3 (IMP3, U3 small nucleolar ribonucleoprotein)
Clonality: monoclonal
Clone: 3B12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR