Anti-IMMT Mouse Monoclonal Antibody [clone: 1A8]

Lieferant: US Biological
USBI128488EA 733 CHF
USBI128488
Anti-IMMT Mouse Monoclonal Antibody [clone: 1A8]
Antikörper
Type: Primary
Antigen: IMMT (inner membrane protein, mitochondrial)
Clonality: monoclonal
Clone: 1A8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR