Anti-ILKAP Mouse Monoclonal Antibody [clone: 3B5]

Supplier: US Biological
USBI128486EA 702 CHF
USBI128486
Anti-ILKAP Mouse Monoclonal Antibody [clone: 3B5]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: ILKAP (integrin-linked kinase-associated serine/threonine phosphatase)
Clonality: monoclonal
Clone: 3B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR