Anti-ILK Mouse Monoclonal Antibody [clone: 4F10]

Supplier: US Biological
USBI128484EA 702 CHF
USBI128484
Anti-ILK Mouse Monoclonal Antibody [clone: 4F10]
Antibodies
Type: Primary
Antigen: ILK (integrin-linked kinase)
Clonality: monoclonal
Clone: 4F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR