Anti-ILF2 Mouse Monoclonal Antibody [clone: 1E2]

Supplier: US Biological
USBI128481EA 702 CHF
USBI128481
Anti-ILF2 Mouse Monoclonal Antibody [clone: 1E2]
Antibodies
Type: Primary
Antigen: ILF2 (interleukin enhancer binding factor 2)
Clonality: monoclonal
Clone: 100
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR