Anti-IL6ST Mouse Monoclonal Antibody [clone: 4A4]

Lieferant: US Biological
USBI128477EA 733 CHF
USBI128477
Anti-IL6ST Mouse Monoclonal Antibody [clone: 4A4]
Antikörper
Type: Primary
Antigen: IL6ST (interleukin 6 signal transducer (gp130, oncostatin M receptor))
Clonality: monoclonal
Clone: 4A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR