Anti-IL5RA Mouse Polyclonal Antibody

Lieferant: US Biological
USBI128473EA 733 CHF
USBI128473
Anti-IL5RA Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL5RA (interleukin 5 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR