Anti-IL4R Mouse Monoclonal Antibody [clone: 1D3]

Supplier: US Biological
USBI128472EA 702 CHF
USBI128472
Anti-IL4R Mouse Monoclonal Antibody [clone: 1D3]
Antibodies
Type: Primary
Antigen: IL4R (interleukin 4 receptor)
Clonality: monoclonal
Clone: 1D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR