Anti-IL4 Mouse Monoclonal Antibody [clone: 3D11]

Supplier: US Biological
USBI128469EA 702 CHF
USBI128469
Anti-IL4 Mouse Monoclonal Antibody [clone: 3D11]
Antibodies
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: monoclonal
Clone: 3D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR