Anti-IL33 Mouse Monoclonal Antibody [clone: 2D8]

Lieferant: US Biological
USBI128467EA 733 CHF
USBI128467
Anti-IL33 Mouse Monoclonal Antibody [clone: 2D8]
Antikörper
Type: Primary
Antigen: IL33 (interleukin 33)
Clonality: monoclonal
Clone: 2D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR