Anti-IL3 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128463EA 788 CHF
USBI128463
Anti-IL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL3 (interleukin 3 (colony-stimulating factor, multiple))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR