Anti-IL3 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128462EA 702 CHF
USBI128462
Anti-IL3 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL3 (interleukin 3 (colony-stimulating factor, multiple))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR