Anti-IL27RA Mouse Monoclonal Antibody [clone: 8G9]

Supplier: US Biological
USBI128457EA 702 CHF
USBI128457
Anti-IL27RA Mouse Monoclonal Antibody [clone: 8G9]
Antibodies
Type: Primary
Antigen: IL27RA (interleukin 27 receptor, alpha)
Clonality: monoclonal
Clone: 8G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR