Anti-IL23R Mouse Monoclonal Antibody [clone: 3D7]

Supplier: US Biological
USBI128452EA 702 CHF
USBI128452
Anti-IL23R Mouse Monoclonal Antibody [clone: 3D7]
Antibodies
Type: Primary
Antigen: IL23R (interleukin 23 receptor)
Clonality: monoclonal
Clone: 3D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR