Anti-IL22RA1 Mouse Monoclonal Antibody [clone: 6B5]

Lieferant: US Biological
USBI128448EA 733 CHF
USBI128448
Anti-IL22RA1 Mouse Monoclonal Antibody [clone: 6B5]
Antikörper
Type: Primary
Antigen: IL22RA1 (interleukin 22 receptor, alpha 1)
Clonality: monoclonal
Clone: 6B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR