Anti-IL2 Mouse Monoclonal Antibody [clone: 1C5]

Supplier: US Biological
USBI128442EA 702 CHF
USBI128442
Anti-IL2 Mouse Monoclonal Antibody [clone: 1C5]
Antibodies
Type: Primary
Antigen: IL2 (interleukin 2)
Clonality: monoclonal
Clone: 1C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR