Anti-IL1RN Mouse Monoclonal Antibody [clone: M1-B9]
Lieferant: US Biological
USBI128441EA 733 CHF
USBI128441
Anti-IL1RN Mouse Monoclonal Antibody [clone: M1-B9]
Antikörper
Type: Primary
Antigen: IL1RN (interleukin 1 receptor antagonist)
Clonality: monoclonal
Clone: M1-B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human