Anti-IL1RAP Mouse Monoclonal Antibody [clone: 2A3]

Supplier: US Biological
USBI128437EA 702 CHF
USBI128437
Anti-IL1RAP Mouse Monoclonal Antibody [clone: 2A3]
Antibodies
Type: Primary
Antigen: IL1RAP (interleukin 1 receptor accessory protein)
Clonality: monoclonal
Clone: 2A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR